Läs mer
Tocris Bioscience™ MK6-83
TRPML channel activator
Brand: Tocris Bioscience™ 5547/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MK6-83 is a TRPML channel activator (EC50 = 110 nM). Also activates F408Δ and F465L TRPML mutants (EC50 values are 0.1 and 1.23 μM respectively). Restores endolysosomal trafficking and zinc homeostasis in lysosomes of mucolipidosis type IV (MLIV) mutant fibroblasts.Specifikationer
| MK6-83 | |
| 5-Methyl-N-[2-(1-piperidinyl)phenyl]-2-thiophenesulfonamide | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| Bioactive Small Molecule |
| 1062271-24-2 | |
| 50 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use