missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MKNK2 (Human) Recombinant Protein
Human MKNK2 full-length ORF ( AAH18345, 1 a.a. - 158 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002872-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAPEVVEAFSEEASIYDKRCDLWSLGVILYILLSGYPPFVGRCGSDCGWDRGEACPACQNMLFESIQEGKYEFPDKDWAHISCAAKDLISKLLVRDAKQRLSAAQVLQHPWVQGCAPENTLPTPMVLQRWDSHFLLPPHPCRIHVRPGGLVRTVTVNE
Specifications
AAH18345 | |
MAP kinase interacting serine/threonine kinase 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MKNK2 | |
GST |
2872 | |
Wheat germ expression system | |
10 μg | |
GPRK7, MNK2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |