missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKP-2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1G1C12, Proteintech
Mouse Monoclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | MKP-2 |
---|---|
Clone | 1G1C12 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828145
|
Proteintech
66349-1-IG-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818145
|
Proteintech
66349-1-IG-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DUSP4 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. DUSP4 inactivates ERK1, ERK2 and JNK, is expressed in a variety of tissues, and is localized in the nucleus.Specifications
MKP-2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
dual specificity phosphatase 4, DUSP4, HVH2, MAP kinase phosphatase 2, MKP 2, MKP2, MKP-2, TYP, VH2 | |
Dusp4 | |
IgG1 | |
Protein G | |
Antibody |
1G1C12 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Rat, Human, Mouse, Porcine | |
Q13115, Q62767, Q8BFV3 | |
100514374, 1846, 319520, 60587 | |
MKP-2 Fusion Protein Ag1206 | |
Primary | |
-20°C | |
DUSP4 |