Läs mer
Tocris Bioscience™ MKT 077
Produktkod. 18074012
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18074012 10 mg
30001679 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18074012 Leverantör Tocris Bioscience™ Leverantörsnummer 4621/10
Binds mot-2; selectively cytotoxic in cancer cells
Occupies mortalin-2 (mot-2), a member of the Hsp70 family, at its p53 binding site and enables p53 translocation to the nucleus. Selectively cytotoxic; causes growth arrest of cancer cells in culture. Also inhibits telomerase activity and cross-links F-actin.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147366-41-4
Molekylformel C21H22ClN3OS2
Specifikationer
Kemiskt namn eller material MKT 077
Kvantitet 10 mg
Formel vikt 432
Renhet >98%
Molekylvikt (g/mol) 432.00