Learn More
ML-098, MedChemExpress
Product Code. 16450378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450378 5 mg 5mg
16443696 25 mg 25mg
16407918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16450378 Supplier Medchem Express Supplier No. HY19800
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress ML-098 (CID-7345532) is an activator of the GTP-binding protein Rab7 with an EC50 of 77.6 nM.
ML-098 (CID-7345532) is an activator of the GTP-binding protein Rab7 with an EC50 of 77.6 nM.
Specifications
Chemical Name or Material ML-098
CAS 878978-76-8
Color White
Purity Grade Notes Research
Molecular Formula C19H19NO3
Quantity 5 mg
Synonym CID-7345532
Solubility Information DMSO : ≥ 29 mg/mL (93.74 mM)
SMILES O=C(C1=CN(CCOC2=CC(C)=CC=C2C)C3=C1C=CC=C3)O
Molecular Weight (g/mol) 309.36
Formula Weight 309.36
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease