Läs mer
Tocris Bioscience™ ML 179
Selective liver receptor homolog 1 (LRH1) inverse agonist
Brand: Tocris Bioscience™ 4957/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ML 179 is a selective liver receptor homolog 1 (LRH1, NR5A2) inverse agonist (IC50 = 320 nM and maximum efficacy of 64% repression). Exhibits >31-fold selectivity over SF1.Specifikationer
| ML 179 | |
| 3-Cyclohexyl-6-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]-1H-pyrimidine-2,4-dione | |
| Soluble to 50mM in DMSO | |
| Bioactive Small Molecule |
| 1883548-87-5 | |
| 10 mg | |
| Crystalline Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use