Läs mer
Tocris Bioscience™ ML 210
Beskrivning
ML 210 is a glutathione peroxidase (GPX4) inhibitor. Kills mutant RAS-expressing cell lines (IC50 values are 71 and 272 nM for HRAS G12v mutant expressing cell lines BJeLR and BJeH-LT, respectively). Exhibits 4-fold selectivity for HRAS mutant-expressing cell lines. Induces ferroptosis in tumor initiation "persister" cells and drug-resistant tumor cells.
Specifikationer
Specifikationer
| Kemiskt namn eller material | [4-[Bis(4-chlorophenyl)methyl]-1-piperazinyl](5-methyl-4-nitro-3-isoxazolyl)methanone |
| CAS | 1360705-96-9 |
| Kvantitet | 10 mg |
| Molekylformel | C22H20Cl2N4O4 |
| Formel vikt | 475.32 |
| Renhet | 98% |