Läs mer
ML 323, Tocris Bioscience™
GREENER_CHOICE
Potent and selective USP1-UAF1 allosteric inhibitor
Brand:  Tocris Bioscience 6435/25
Encompass
Produktkod. 15977940
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
ML 323 is a potent and selective allosteric inhibitor of USP1-UAF1 deubiquitinase complex (IC50 = 76 nM). Exhibits selectivity for USP1-UAF1 over a panel of enzymes, including other deuquitinating enzymes. Inhibits deubiquitination of PCNA and FANCD2 and sensitizes cisplatin-resistant H596 cells to cell death induced by cisplatin (Cat. No. 2251) in vitro.
Specifikationer
Specifikationer
1572414-83-5
5-Methyl-2-[2-(1-methylethyl)phenyl]-N-[[4-(1H-1,2,3-triazol-1-yl)phenyl]methyl]-4-pyrimidinamine