Läs mer
Tocris Bioscience™ ML 337
Kvantitet:
10 mg
50 mg
Beskrivning
ML 337 is a selective negative allosteric modulator of mGlu3 (IC50 = 593 nM). Displays no activity at mGlu1, mGlu2 or mGlu4-8 at concentrations up to 30 μM. Brain penetrant.
Specifikationer
Specifikationer
| Kvantitet | 10 mg |
| Produkttyp | ML 337 |
| Renhet | >98% |
| Molekylvikt (g/mol) | 353.39 |
| Inhibitorer | ML 337 |
For Research Use Only