Läs mer
Tocris Bioscience™ ML 337
Selective negative allosteric modulator of mGlu3
Brand:  Tocris Bioscience™ 4982/10
Encompass
Produktkod. 15746366
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
ML 337 is a selective negative allosteric modulator of mGlu3 (IC50 = 593 nM). Displays no activity at mGlu1, mGlu2 or mGlu4-8 at concentrations up to 30 μM. Brain penetrant.
Specifikationer
Specifikationer
>98%
10 mg
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer