Läs mer
Tocris Bioscience™ ML 339
Potent and selective hCXCR6 antagonist
Brand: Tocris Bioscience™ 5943/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ML 339 is a potent and selective hCXCR6 antagonist (IC50 = 140 nM); 100-fold less active at the murine CXCR6 receptor (IC50 = 18 μM). Exhibits selectivity over CXCR5, CXCR4, CCR6 and APJ receptors (IC50 >79 μM).Specifikationer
| N-[(1 R,3 S,5 S)-9-(2-((2-Chlorophenyl)amino)-2-exoethyl)-9-azabicyclo[3.3.1 ]nonan-3-yl)-3,4,5-trimethoxybenzamide | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
| 2080300-49-6 | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| White |