Läs mer
Tocris Bioscience™ ML 382
Beskrivning
ML 382 is a selective MRGPRX1 positive allosteric modulator (EC50 = 190 nM). Exhibits selectivity for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters. Enhances BAM (8-22) potency at MRGPRX1 by 7-fold.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Mas-Related G Protein Coupled Receptor Modulators |
| CAS | 1646499-97-9 |
| Kvantitet | 50 mg |
| Mål | Sensory Neuron-Specific Receptor Agonists |
| Molekylformel | C18H20N2O4S |
| Formel vikt | 360.43 |
| Renhet | >98% |