Läs mer
ML224, MedChemExpress
Produktkod. 16497847
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497847 5 mg -
16495128 10 mg -
16485846 100 mg 100mg
16441367 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16497847 Leverantör Medchem Express Leverantörsnummer HY12381
MedChemExpress ML224(NCGC00242364; ANTAG3) is a selective TSHR inverse agonist; inhibits TSH-stimulated cAMP production with an IC50 = 2.3 μM.
ML224(NCGC00242364
Specifikationer
Kemiskt namn eller material ML224
CAS 1338824-21-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C31H31N3O5
Kvantitet 5 mg
Synonym NCGC00242364 ANTAG3
Löslighetsinformation DMSO : 100 mg/mL (190.26 mM; Need ultrasonic)
LEDER CC(NC1=CC(C)=C(OCC2=CC(C(N3CC4=CC=CO4)NC5=C(C=CC=C5)C3=O)=CC=C2OC)C(C)=C1)=O
Molekylvikt (g/mol) 525.59
Formel vikt 525.59
Procent renhet 99.71%
Kvalitet Research
Fysisk form Solid