Läs mer
ML228, MedChemExpress
Produktkod. 16475318
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16475318 10 mg
16465966 100 mg
16428037 5 mg
16421527 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16475318 Leverantör Medchem Express Leverantörsnummer HY12754
MedChemExpress ML228 (CID-46742353) is a potent the Hypoxia Inducible Factor (HIF) pathway activator with EC50 of 1 μM. ML228 potently activates HIF in vitro as well as its downstream target VEGF.
ML228 (CID-46742353) is a potent the Hypoxia Inducible Factor (HIF) pathway activator with EC50 of 1 μM. ML228 potently activates HIF in vitro as well as its downstream target VEGF.
Specifikationer
Kemiskt namn eller material ML228
Fysisk form Solid
CAS 1357171-62-0
Färg Light Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C27H21N5
För användning med (applikation) Cancer-programmed cell death
Synonym CID-46742353
Löslighetsinformation DMSO : ≥ 35 mg/mL (84.24 mM)
LEDER C1(C2=NC=CC=C2)=NC(NCC3=CC=C(C4=CC=CC=C4)C=C3)=C(C5=CC=CC=C5)N=N1
Molekylvikt (g/mol) 415.49
Formel vikt 415.49
Procent renhet 98.0%