missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MLCK (MLCK2), GST Tag Recombinant Protein
Human MLCK (MLCK2), GST Tag Recombinant Protein
18240.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_872299, R175G |
---|---|
Gene Alias | MLCK (MLCK2) |
KinaseFamily | MLCK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428533
|
Invitrogen™
PR7174A |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10568113
|
Invitrogen™
PV4023 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MLCK (MLCK2) is of of the myosin regulatory light chain kinases which facilitates muscle contraction.
Specifications
NP_872299, R175G | |
MLCK Kinase Family | |
Recombinant | |
Baculovirus |
MLCK (MLCK2) | |
Human | |
GST Tag |