missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MLF1IP Recombinant Protein
Brand: Abnova™ H00079682-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH31520 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
45 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MKTSDIKELNIVLPEFEKTHLEHQQRIESKVCKAAIATFYVNVKEQFIKMLKESQMLTNLKRKNAKMISDIEKKRQRMIEVQDELLRLEPQLKQLQTKYDELKERKSSLRNAAYFLSNLKQLYQDYSDVQAQEPNVKETYDSSSLPALLFKARTLLGAESHLRNINHQLEKLLDQG | |
CENP-50/CENP-U/CENP-U(50)/CENP50/CENPU/FLJ23468/KLIP1/PBIP1 | |
MLF1IP | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
79682 | |
MLF1IP (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MLF1IP | |
Human | |
Recombinant | |
Solution |