missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
MLH3 Monoclonal antibody specifically detects MLH3 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MLH3 |
| Användningsområden | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Klassificering | Monoclonal |
| Klona | CL14096 |
| Konjugera | Alexa Fluor 488 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | DNA mismatch repair protein Mlh3, HNPCC7, MGC138372, mutL (E. coli) homolog 3, mutL homolog 3 (E. coli), MutL protein homolog 3 |
| Värd art | Mouse |
| Immunogen | This antibody was generated using a synthetic peptide of Q9UHC1, with the exact immunogen sequence remaining proprietary. |
| Reningsmetod | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?