Learn More
Sartorius mLINE™ Pipette Multipacks
Replenish or set up a new pipetting station. Sartorius mLINE™ Pipette Multipacks conveniently combine a set of mLINE mechanical pipettes with matching tip trays and useful accessories. Available in collections of 3, 4 and 5 pipettes with matching tips
Brand: Sartorius LH-725662
Additional Details : Weight : 2.60000kg
Description
Multipacks to match every need- Affordable options to get a set of quality pipettes with useful accessories
- Ideal for renewing an existing pipette collection or setting up a new pipetting station
- Includes a linear pipette rack, calibration tool, tube opener and color-coding caps
- Collections include up to five different volumes of single channel pipettes, from 10μL to 5mL; the 3+1 pack includes a 350μL eight-channel pipette
- Offer excellent ergonomics, performance, precision and safety in manual pipetting
- Requires exceptionally light pipetting and tip ejection forces
- Specifically designed to prevent Repetitive Strain Injury (RSI) in repetitive pipetting
- Minimized risk of contamination with Safe-Cone Filters and full autoclavability
- Increased safety with volume lock, preventing accidental volume changes while pipetting
- Designed for the best possible fitting and sealing for the highest possible accuracy and precision
- Tips are RNase, DNase and endotoxin free and compliant with ISO 9001 and ISO 13485
- Environment-friendly materials and fully autoclavable to reduce waste
Specifications
Combines mLINE pipets and Optifit Tips with for safe, precise pipetting | |
Repetitive Manual Pipetting | |
mLINE | |
Repetitive manual pipetting | |
3 pipettes with 3 boxes of Optifit Trays |
Autoclavable | |
1 | |
Pipette Package | |
Pipets: 20μL, 200μL and 1,000μL, single channel Optifit Tip tray boxes: 2 x 200μL and 1,000μL Linear stand and accessories |