Learn More
MLK4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552460
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GNPTPTGATI ISATGASALP LCPSPAPHSH LPREVSPKKH STVHIVPQRR PASLRSRSDL PQAYPQTAVS QLAQTACVVG RPGPHPTQFL AAKERTKSHV PSLLDADVEG QSRDYTVPLC RMRSKTSRPS IYELEKE Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 34%.
MLK4 Polyclonal antibody specifically detects MLK4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MLK4 | |
Polyclonal | |
Unconjugated | |
MAP3K21 | |
BC021891; cDNA sequence BC021891; dJ862P8.3; Kiaa1804; MAP3K21; Mitogen-activated protein kinase kinase kinase 21; Mitogen-activated protein kinase kinase kinase MLK4; mixed lineage kinase 4; mKIAA1804; MLK4; RGD1306091; similar to Mixed lineage kinase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TCX8 | |
MAP3K21 | |
Recombinant protein corresponding to Human MLK4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only