missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549581
Additional Details : Weight : 0.01000kg
Description
KMT2D Polyclonal antibody specifically detects KMT2D in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
KMT2D | |
Polyclonal | |
Unconjugated | |
KMT2D | |
AAD10; ALL1-related protein; ALR; CAGL114; Histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; KABUK1; Kabuki make-up syndrome; Kabuki mental retardation syndrome; KMS; KMT2D; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2D; lysine N-methyltransferase 2D; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia 2; Myeloid/lymphoid or mixed-lineage leukemia protein 2; TNRC21; trinucleotide repeat containing 21 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8085 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O14686 | |
KMT2D | |
KLH conjugated synthetic peptide between 4980-5009 amino acids from the C-terminal region of human MLL2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |