missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MLLT6 (Human) Recombinant Protein
Human MLLT6 full-length ORF ( AAH07237, 1 a.a. - 462 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004302-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGAVNPLLSQAESSHTEPDLEDCSFRCRGTSPQESLSSMSPISSLPALFDQTASAPCGGGQLDPAAPGTTNMEQLLEKQGDGEAGVNIVEMLKALHALQKENQRLQEQILSLTAKKERLQILNVQLSVPFPALPAALPAANGPVPGPYGLPPQAGSSDSLSTSKSPPGKSSLGLDNSLSTSSEDPHSGCPSRSSSSLSFHSTPPPLPLLQQSPATLPLALPGAPAPLPPQPQNGLGRAPGAAGLGAMPMAEGLLGGLAGSGGLPLNGLLGGLNGAAAPNPASLSQAGGAPTLQLPGCLNSLTEQQRHLLQQQEQQLQQLQQLLASPQLTPEHQTVVYQMIQQIQQKRELQRLQMAGGSQLPMASLLAGSSTPLLSAGTPGLLPTASAPPLLPAGALVAPSLGNNTSLMAAAAAAAAVAAAGGPPVLTAQTNPFLSLSGAEGSGGGPKGGTADKGASANQEKG
Specifications
AAH07237 | |
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MLLT6 | |
GST |
4302 | |
Wheat germ expression system | |
10 μg | |
AF17, FLJ23480 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |