Läs mer
Tocris Bioscience™ MLN 4760
Beskrivning
MLN 4760 is a highly potent, cell-permeable and selective inhibitor of angiotensin converting enzyme 2 (ACE2) (IC50 = 0.44 nM). MLN 4760 inhibits production of angiotensin (1-7) from angiotensin II and reduces ACE2 expression and activity in an animal model of lung inflammation.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[(1 S)-1-Carboxy-3-methylbutyl]-3-[(3,5-dichlorophenyl)methyl]-L-histidine |
| CAS | 305335-31-3 |
| Kvantitet | 2 mg |
| Mål | ACE Inhibitors |
| Molekylformel | C19H23Cl2N3O4 |
| Renhet | 0.98 |