missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10371-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22. 3. Alternative splicing results in multiple transcript variants.Specifications
MMP1 | |
Polyclonal | |
Unconjugated | |
MMP1 | |
CLG, CLGN, Fibroblast collagenase, Interstitial collagenase, Matrix metalloproteinase 1, MMP 1, MMP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
300339, 4312, 83995 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P03956, P81563, Q9EPL5 | |
Mmp1, Mmp1a | |
MMP1 Fusion Protein Ag0547 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |