missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Human MMP-9 Recombinant Protein
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
Beskrivning
This product contains purified recombinant human MMP-9 protein with an N-terminal hexahistidine tag. The recombinant protein is a fragment corresponding to the catalytic domain of the protein (residues 113-450). This product may be used as a positive control in many immunological assays.
MMP9 (matrix metallopeptidase 9, GELB, CLG4B) is a matrix metalloproteinase, a family of zinc and calcium-dependent endopeptidases that degrade extracellular matrix proteins. MMP9 is secreted as a 92kDa zymogen and cleavage of pro-MMP9 results in the active enzyme with a molecular weight of 82kDa. MMP9 has a gelatin-binding domain consisting of three fibronectin type II units, a catalytic domain containing the zinc-binding site, a proline-rich type V collagen-homologous domain and a hemopexin-like domain. MMP9 is produced by monocytes, macrophages, neutrophils, keratinocytes, fibroblasts, osteoclasts and endothelial cells, and is involved in inflammatory responses, tissue remodeling, wound healing, tumor growth and metastasis. MMP9 is supplied by bone marrow-derived cells and contributes to skin carcinogenesis. Further, MMP9 degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. Studies have shown that elevated MMP9 is associated with progression of idiopathic atrial fibrillation and aortic aneurysm.
Specifikationer
Specifikationer
| Tillträdesnummer | P14780 |
| Koncentration | 0.27 mg/mL |
| För användning med (applikation) | Control, Western Blot |
| Formulering | 20 mM tris HCl with 50% glycerol and no preservative; pH 8.0 |
| Gen-ID (Entrez) | 4318 |
| Molekylvikt (g/mol) | 42.03 kDa |
| Namn | Human MMP-9 |
| pH-intervall | 8 |
| Kvantitet | 100 μg |
| Källa | E. Coli |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering