missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMS19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16015-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Key component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into apoproteins specifically involved in DNA metabolism and genomic integrity. In the CIA complex, MMS19 acts as an adapter between early-acting CIA components and a subset of cellular target iron-sulfur proteins such as ERCC2/XPD, FANCJ and RTEL1, thereby playing a key role in nucleotide excision repair (NER) and RNA polymerase II (POL II) transcription. As part of the mitotic spindle-associated MMXD complex, plays a role in chromosome segregation, probably by facilitating iron-sulfur cluster assembly into ERCC2/XPD. Indirectly acts as a transcriptional coactivator of estrogen receptor (ER), via its role in iron-sulfur insertion into some component of the TFIIH-machinery.Specifications
MMS19 | |
Polyclonal | |
Unconjugated | |
MMS19 | |
hMMS19, MET18, MET18 homolog, MMS19, MMS19 like protein, MMS19L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171124, 64210, 72199 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96T76, Q9D071 | |
Mms19 | |
MMS19 Fusion Protein Ag8792 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |