Läs mer
MMV008138, MedChemExpress
Produktkod. 16497837
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497837 5 mg -
16475118 10 mg -
16441357 50 mg 50mg
16415846 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16497837 Leverantör Medchem Express Leverantörsnummer HY123561
MedChemExpress MMV008138 is a species-selective IspD (enzyme 2-C-methyl-d-erythritol 4-phosphate cytidylyltransferase)-targeting antimalarial agent, with an IC50 of 44 nM for PfIspD (P. falciparum IspD). MMV008138 inhibits the growth of P. falciparum Dd2 strain with an IC50 of 250 nM.
MMV008138 is a species-selective IspD (enzyme 2-C-methyl-d-erythritol 4-phosphate cytidylyltransferase)-targeting antimalarial agent, with an IC50 of 44 nM for PfIspD (P. falciparum IspD). MMV008138 inhibits the growth of P. falciparum Dd2 strain with an IC50 of 250 nM.
Specifikationer
Kemiskt namn eller material MMV008138
Fysisk form Solid
CAS 1679333-73-3
Färg Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H14Cl2N2O2
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation DMSO : 125 mg/mL (346.05 mM; Need ultrasonic)
LEDER O=C([C@@H]1CC2=C([C@@H](C3=CC=C(Cl)C=C3Cl)N1)NC4=C2C=CC=C4)O
Molekylvikt (g/mol) 361.22
Formel vikt 361.22
Procent renhet 99.48%