Läs mer
Mn(III) meso-Tetra (N-Methyl-4-pyridyl) Porphine Pentachloride, TRC
Produktkod. 30470694
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30470694 500 mg
1 options
Produktkod. 30470694 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM425510500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mn(III) meso-Tetra (N-Methyl-4-pyridyl) Porphine Pentachloride
CAS 125565-45-9
Molekylformel C44H36ClMnN8 . 4Cl
Kvantitet 500 mg
Synonym Chloro[meso-tetrakis(4-n-methylpyridinyl)porphyrinato)manganese(III)tetrachloride, Manganese(III) 5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphin Chloride Tetrakis(methochloride), (SP-5-12)-Chloro[[4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl-kN21,kN22,kN23,kN24)tetrakis[1-methylpyridiniumato]](2-)] Manganese(4+) Tetrachloride, 4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl)tetrakis[1-methylpyridinium Manganese Complex
InChI formel InChI=1S/C44H36N8.ClH.Mn/c1-49-21-13-29(14-22-49)41-33-5-7-35(45-33)42(30-15-23-50(2)24-16-30)37-9-11-39(47-37)44(32-19-27-52(4)28-20-32)40-12-10-38(48-40)43(36-8-6-34(41)46-36)31-17-25-51(3)26-18-31;;/h5-28H,1-4H3;1H;/q+2;;+3/p-1
LEDER C[n+]1ccc(cc1)\C\2=C\3/C=C\C\4=C(\C5=N\C(=C(\c6cc[n+](C)cc6)/c7ccc(\C(=C/8\C=CC2=N8)\c9cc[n+](C)cc9)n7[Mn](Cl)N34)\C=C5)/c%10cc[n+](C)cc%10
Molekylvikt (g/mol) 909
Formel vikt 766.2132