missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ MNAR Monoclonal Antibody (4F1D1)
Beskrivning
MA5-31837 has been tested in indirect ELISA.
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
Specifikationer
Specifikationer
| Antigen | MNAR |
| Användningsområden | ELISA, Flow Cytometry, Western Blot |
| Klassificering | Monoclonal |
| Klona | 4F1D1 |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 0.05% sodium azide |
| Gen | PELP1 |
| Genaccessionsnr. | Q8IZL8 |
| Gene Alias | 4930563C04Rik; HMX3; Mnar; modulator of nongenomic activity of estrogen receptor; Modulator of non-genomic activity of estrogen receptor; P160; PELP1; proline and glutamic acid rich nuclear protein; proline, glutamate and leucine rich protein 1; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; RGD1306320; transcription factor HMX3 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?