missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23742-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The Myc/Max/Mad network comprises a group of transcription factors that co-interact to regulate gene-specific transcriptional activation or repression. This gene encodes a protein member of the Myc/Max/Mad network. This protein has a basic-Helix-Loop-Helix-zipper domain (bHLHzip) with which it binds the canonical DNA sequence CANNTG, known as the E box, following heterodimerization with Max proteins. This protein is likely a transcriptional repressor and an antagonist of Myc-dependent transcriptional activation and cell growth. This protein represses transcription by binding to DNA binding proteins at its N-terminal Sin3-interaction domain.Specifications
MNT | |
Polyclonal | |
Unconjugated | |
MNT | |
bHLHd3, MAD6, MAX binding protein, Max binding protein MNT, MNT, MXD6, Myc antagonist MNT, Protein ROX, ROX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4335 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99583 | |
MNT | |
MNT Fusion Protein Ag20666 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |