Läs mer
Tocris Bioscience™ MO-I-500
Beskrivning
MO-I-500 is a FTO inhibitor (IC50 = 8.7 μM for purified FTO in vitro), which increases N6-methyladenosine (m6A) levels in total cellular mRNA in HeLa cells. MO-I-500 modulates levels of specific miRNAs. In a triple-negative breast cancer cell line metabolically challenged by culture without glutamine (SUM149-MA cells), MO-I-500 inhibits cell proliferation. MO-I-500 also displays anticonvulsant activity in a 6 Hz mouse model of epilepsy at nontoxic doses.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[5-(4-Chlorophenyl)-3,4-dihydroxy-2-furanyl]ethanesulfonamide |
| CAS | 1585219-04-0 |
| Kvantitet | 50 mg |
| Mål | Other Oxidase/Oxygenase Compounds |
| Molekylformel | C12H12ClNO5S |
| Renhet | 0.98 |