Learn More
MOBS, 0.2M buffer soln., pH 7.0, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J61416.AK
Additional Details : CAS Number : 115724-21-5 Weight : 0.00025kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
115724-21-5 | |
0.2 M | |
Liquid | |
C8H17NO4S | |
4-(4-Morpholinyl)butanesulfonic acid | |
OS(=O)(=O)CCCCN1CCOCC1 | |
223.29 |
Colorless | |
7.0 | |
250 mL | |
MFCD00674916 | |
VTOWJTPBPWTSMK-UHFFFAOYSA-N | |
4-(morpholin-4-yl)butane-1-sulfonic acid |