Läs mer
Modafinil Carboxylate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Modafinil acid |
| CAS | 63547-24-0 |
| Molekylformel | C15 H14 O3 S |
| Kvantitet | 50 mg |
| Synonym | Acetic acid, 2-[(diphenylmethyl)sulfinyl]-, Acetic acid, [(diphenylmethyl)sulfinyl]- (9CI), 2-[(Diphenylmethyl)sulfinyl]acetic acid, (Benzhydrylsulfinyl)acetic acid, (±)-Modafinic acid, CRL 40467, Modafinil acid, Modafinilic acid, [(RS)-(Diphenylmethyl)sulfinyl]acetic acid, Modafinil carboxylate |
| InChI formel | InChI=1S/C15H14O3S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H,16,17) |
| LEDER | OC(=O)CS(=O)C(c1ccccc1)c2ccccc2 |
| IUPAC-namn | 2-benzhydrylsulfinylacetic acid |
| Molekylvikt (g/mol) | 274.33 |
| Formel vikt | 274.0664 |