Learn More
Hamilton™ 1700 Series Removable Needle Syringes
Ideal for dispensing both liquids and gases.
Brand:  Hamilton™ DS80065
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11528024
  • 19795.00 SEK / Each
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
  • Precision-machined PTFE plunger tip which creates a leak-free seal
  • Tip wipes the interior of the syringe barrel free of sample
  • Fluid path is chemically inert as possible
Specifications
Specifications
22s G
10 μL