Learn More
Hamilton™ Thermo Finnagan™ Cemented Needle Syringes
Enables the automatic introduction of samples into the injector of the gas chromatograph.
Brand:  Hamilton™ 204000
Additional Details : Weight : 0.02400kg
Product Code. 11547614
  • 1280.00 SEK / Each
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
  • Improves reproducibility and speed
  • Point style specifically designed to withstand repeated penetration through the GC septa
  • For use with Thermo Finnigan™ AI/AS 3000 TriPlus™ autosamplers
Specifications
Specifications
5 μL