Learn More
Asecos™ Module For Remote Signaling 4.0 ION PRO And ULTRA
Product Code. 31789613 Shop All asecos Products
Change view
Click to view available options
Model:
EU-Version
Unit Size:
Each
Product Code. Model Unit Size
31789613 EU-Version Each
1 options
Product Code. 31789613 Supplier Asecos™ Supplier No. 42483
 Restricted Item
Beräknad leverans: 14-12-2026
Remote signaling module enables remote alarm forwarding via mobile networks (SMS/voice)
  • Module for remote signalling 4.0 ION-LINE EU/UK
    • Forwards all four alarm statuses (warning message, alarm level 1, alarm level 2, power failure) in a differentiated manner via SMS and/or call to the mobile network.
    • Alarm texts and up to 5 phone numbers are freely configurable.
  • Prerequisites
    • Network availability at the installation site (4G LTE-M and 2G). If you have your own SIM card, the module only uses 2G the SIM must be provided by the customer.
    • Alternatively, a Mobeye SIM is included, which is activated via an online portal (extended contact options via SMS and e-mail, costs according to Mobeye). 230 V mains connection required.
    • Compatible with IO90.195.120.065.WDEL
  • Special feature
    • For models IO90.195.120.PC.WDC, IO90.195.120.PS.WDC only compatible in combination with relay module 4.0 [41405].
    • Can be retrofitted by the customer.
Specifications
For Use With (Application) Forwarding of Alarm Conditions
For Use With (Equipment) 4.0 ION PRO and ULTRA, EU
Model EU-Version
Weight (English) 1.10 lb.
Weight (Metric) 0.5 kg
Product Type Remote Signaling Module