Läs mer
Miele Professional™ Mobile Injector, Module A 306
Produktkod. 15455021 Handla allt Miele Produkter
:
for 4 1-2 litre measuring cylinders, high shape; model: glassware
Produktkod. 15455021
Brand: Miele Professional™ 9862580
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Module A 306 for the optimum loading of large volume laboratory glassware
  • Suitable for 4 1-2 litre measuring cylinders, high shape
  • Suitable for 2 1-2 litre measuring cyl., low shape + 2 high shape
  • Contact surfaces plastic coated