Läs mer
Moexipril Cyclohexyl Analogue Hydrochloride, TRC
Produktkod. 30479819
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479819 5 mg
30479097 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479819 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM4853355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1356019-89-0
Molekylformel C27 H40 N2 O7 . H Cl
Molekylvikt (g/mol) 541.077
Synonym (S)-2-{(S)-2-[(S)-4-Cyclohexyl-1-ethoxy-1-oxobutan-2-ylamino]propanoyl}-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid Hydrochloride
IUPAC-namn (3S)-2-[(2S)-2-[[(2S)-4-cyclohexyl-1-ethoxy-1-oxobutan-2-yl]amino]propanoyl]-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
LEDER Cl.CCOC(=O)[C@H](CCC1CCCCC1)N[C@@H](C)C(=O)N2Cc3cc(OC)c(OC)cc3C[C@H]2C(=O)O
Specifikationer
Kemiskt namn eller material Moexipril Cyclohexyl Analogue Hydrochloride
Kvantitet 5 mg
InChI formel InChI=1S/C27H40N2O7.ClH/c1-5-36-27(33)21(12-11-18-9-7-6-8-10-18)28-17(2)25(30)29-16-20-15-24(35-4)23(34-3)14-19(20)13-22(29)26(31)32;/h14-15,17-18,21-22,28H,5-13,16H2,1-4H3,(H,31,32);1H/t17-,21-,22-;/m0./s1
Formel vikt 540.26