Läs mer
Moexipril Methyl Ester Analog Hydrochloride Salt, TRC
Produktkod. 30471914
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30471914 10 mg
1 options
Produktkod. 30471914 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM48534510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Moexipril Methyl Ester Analog Hydrochloride Salt
CAS 1356841-17-2
Molekylformel C26 H32 N2 O7 . H Cl
Kvantitet 10 mg
Synonym [3S-[2-(S),3S]]-1,2,3,4-Tetrahydro-6,7-dimethoxy-2-[2-[[1-(methoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-3-isoquinolinecarboxylic Acid Hydrochloride
InChI formel InChI=1S/C26H32N2O7.ClH/c1-16(27-20(26(32)35-4)11-10-17-8-6-5-7-9-17)24(29)28-15-19-14-23(34-3)22(33-2)13-18(19)12-21(28)25(30)31;/h5-9,13-14,16,20-21,27H,10-12,15H2,1-4H3,(H,30,31);1H/t16-,20-,21-;/m0./s1
LEDER Cl.COC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2Cc3cc(OC)c(OC)cc3C[C@H]2C(=O)O
IUPAC-namn (3S)-6,7-dimethoxy-2-[(2S)-2-[[(2S)-1-methoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 521.002
Formel vikt 520.198