Läs mer
Moexiprilat Hydrochloride, TRC
Produktkod. 30476845
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30476845 5 mg
1 options
Produktkod. 30476845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM4853605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Moexiprilat hydrochloride
CAS 82586-57-0
Molekylformel C25 H30 N2 O7 . Cl H
Kvantitet 5 mg
Synonym 3-Isoquinolinecarboxylic acid, 2-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-, monohydrochloride, (3S)- (9CI), 3-Isoquinolinecarboxylic acid, 2-[2-[(1-carboxy-3-phenylpropyl)amino]-1-oxopropyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-, monohydrochloride, [3S-[2[R*(R*)],3R*]]-
InChI formel InChI=1S/C25H30N2O7.ClH/c1-15(26-19(24(29)30)10-9-16-7-5-4-6-8-16)23(28)27-14-18-13-22(34-3)21(33-2)12-17(18)11-20(27)25(31)32;/h4-8,12-13,15,19-20,26H,9-11,14H2,1-3H3,(H,29,30)(H,31,32);1H/t15-,19-,20-;/m0./s1
LEDER Cl.COc1cc2C[C@H](N(Cc2cc1OC)C(=O)[C@H](C)N[C@@H](CCc3ccccc3)C(=O)O)C(=O)O
IUPAC-namn (3S)-2-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 506.9758
Formel vikt 506.182