Läs mer
Mofebutazone, TRC
Produktkod. 30004604
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30004604 250 mg
1 options
Produktkod. 30004604 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM485400250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mofebutazone
CAS 2210-63-1
Molekylformel C13 H16 N2 O2
Kvantitet 250 mg
Synonym 3,5-Pyrazolidinedione, 4-butyl-1-phenyl-, 4-Butyl-1-phenyl-3,5-pyrazolidinedione, 1-Phenyl-4-butyl-3,5-pyrazolidinedione, 1-Phenyl-4-butylpyrazolidin-3,5-dione, 1-Phenyl-4-n-butyl-3,5-dioxopyrazolidine, 2-Phenyl-3,5-dihydroxy-4-butylpyrazolidine, 4-Butyl-1-phenyl-3,5-dioxopyrazolidine, 4-Butyl-3,5-dioxo-1-phenyltetrahydropyrazole, Arcobutine, Arcomonol Tablets, Butazone, Mobutazon, Mobutazone, Mobuzon, Mofebutazone, Mofesal, Monazan, Monazon, Monobutyl, Monofen, Monofenew, Monomil, Monophenylbutazone, Monorheumetten, Monozon, Mophebutazone, Mozol, NSC 73725, Reumatox
InChI formel InChI=1S/C13H16N2O2/c1-2-3-9-11-12(16)14-15(13(11)17)10-7-5-4-6-8-10/h4-8,11H,2-3,9H2,1H3,(H,14,16)
LEDER CCCCC1C(=O)NN(C1=O)c2ccccc2
IUPAC-namn 4-butyl-1-phenylpyrazolidine-3,5-dione
Molekylvikt (g/mol) 232.28
Formel vikt 232.1212