Läs mer
Moldex™ 8000 Series Half-Mask Facepieces
Formatera:
Small
Medium
Förpackningsstorlek:
Varje
Beskrivning
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
- Extra wide surface seals easily and stays sealed even when talking
- Low profile design provides a large field of vision
- Facepiece has no gaskets or yoke; fewer parts to maintain and inventory
- Each cartridge includes a new inhalation valve and features snap-in mount that eliminates mis-threading
Specifikationer
Specifikationer
| Certifikat och efterlevnad | NIOSH |
| Anslutningstyp | Snap-in gasket |
| Material | Thermoplastic Elastomer |
| Produkttyp | Half Mask Facepiece |
| Formatera | Medium |