Läs mer
Invitrogen™ D-Myo-Inositol 1,4,5-Triphosphate, P4(5)-(1-(2-Nitrophenyl)ethyl) Ester, tris(triethylammonium) Salt (NPE-Caged Ins 1,4,5-P3)
Produktkod. 11544896
Change view
Kvantitet:
25 μg
Förpackningsstorlek:
25µg
Produktkod. Kvantitet unitSize
11544896 25 μg 25µg
1 options
Produktkod. 11544896 Leverantör Invitrogen™ Leverantörsnummer I23580
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Flash photolysis of the caging group yields highly localized release of the free nucleotide
NPE-caged inositol 1,4,5-triphosphate is biologically inactive before photolysis. Flash photolysis of the caging group with UV illumination at 355nm results in a rapid and highly localized release of free INS 1,4,5-P3 at the site of illumination.
Cell Analysis, Cell Structure, Cellular Imaging, Enzyme and Protein Activity Assays, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis
Beställningsinfo
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 25 μg