Läs mer
Invitrogen™ Molecular Probes™ Merocyanine 540
Beskrivning
- Binds to surface of polarized membranes in perpendicular orientation, reorienting as membrane depolarizes to form nonfluorescent dimers with altered absorption spectra
- Used as photosensitizer
Cell Analysis, Cell Metabolism, Cell Structure, Cell Viability, Proliferation and Function, Membranes (General) and Lipids, Nitro-Oxidative Stress, Reactive Oxygen Species
Beställningsinfo
Shipping Condition: Room temperature
Specifikationer
Specifikationer
| Färg | Yellow |
| Innehåll och lagring | Store at Room temperature and protect from light. |
| För användning med (utrustning) | Fluorescence Microscope |
| Kvantitet | 25 mg |