Läs mer
Molybdenum Powder, Sigma-Aldrich™
Beskrivning
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. Molybdenum disulfide (MoS2) is used as a catalyst for hydrodesulfurization in petroleum refining when supported on alumina, enabling efficient removal of sulfur compounds, offering a greener, potentially more sustainable option for fuel processing and environmental protection.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Molybdenum Powder |
| CAS | 7439-98-7 |
| Smältpunkt | 2620°C |
| Densitet | 10.2 g/cm3 at 20°C |
| Kvantitet | 100 g |
| Form | Powder |
| MDL-nummer | MFCD00003465 |
| Synonym | Molybdenum powder, Molybdenum powder |
| Molekylvikt (g/mol) | 95.94 |