Läs mer
Momelotinib Benzoic Acid, TRC
Produktkod. 30470995
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30470995 25 mg
1 options
Produktkod. 30470995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM96562025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Momelotinib Benzoic Acid
CAS 945749-71-3
Molekylformel C21H20N4O3
Kvantitet 25 mg
Synonym 4-[2-[[4-(4-Morpholinyl)phenyl]amino]-4-pyrimidinyl]benzoic acid, 4-[2-(4-Morpholinophenylamino)pyrimidin-4-yl]benzoic acid
InChI formel InChI=1S/C21H20N4O3/c26-20(27)16-3-1-15(2-4-16)19-9-10-22-21(24-19)23-17-5-7-18(8-6-17)25-11-13-28-14-12-25/h1-10H,11-14H2,(H,26,27)(H,22,23,24)
LEDER O=C(O)C1=CC=C(C2=NC(NC3=CC=C(N4CCOCC4)C=C3)=NC=C2)C=C1
Molekylvikt (g/mol) 376.42
Formel vikt 376.15