Läs mer
Mometasone Furoate Impurity A, TRC
Produktkod. 30138346
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138346 10 mg -
30135344 25 mg -
17980745 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM49010010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83880-65-3
Molekylformel C27 H29 Cl O5
Molekylvikt (g/mol) 468.97
Synonym Pregna-1,4,9(11)-triene-3,20-dione, 21-chloro-17-[(2-furanylcarbonyl)oxy]-16-methyl-, (16α)-, 21-Chloro-16α-methyl-3,20-dioxopregna-1,4,9(11)-trien-17-yl Furan-2-carboxylate, Mometasone Furoate Imp. A (EP)
IUPAC-namn [(8S,10S,13S,14S,16R,17R)-17-(2-chloroacetyl)-10,13,16-trimethyl-3-oxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl
Specifikationer
Kemiskt namn eller material 21-Chloro-16alpha-methyl-3,20-dioxopregna-1,4,9(11)-trien-17-yl Furan-2-carboxylate
Kvantitet 10 mg
InChI formel InChI=1S/C27H29ClO5/c1-16-13-21-19-7-6-17-14-18(29)8-10-25(17,2)20(19)9-11-26(21,3)27(16,23(30)15-28)33-24(31)22-5-4-12-32-22/h4-5,8-10,12,14,16,19,21H,6-7,11,13,15H2,1-3H3/t16-,19-,21+,25+,26+,27+/m1/s1
Formel vikt 468.1704