Learn More
Thermo Scientific Alfa Aesar Monel|r 400 gauze, 150 mesh woven from 0.066mm (0.0026in) dia wire 300x300mm
Brand: Thermo Scientific Alfa Aesar 045185.HB
Monel|r 400 gauze, 150 mesh woven from 0.066mm (0.0026in) dia wire 300x300mm
Additional Details : Weight : 0.01000kg