Learn More
Alfa Aesar™ Monel™ 400 gauze, 150 mesh woven from 0.066mm (0.0026 in.) dia. wire
Brand:  Alfa Aesar™ 045185.GT
Additional Details : Weight : 0.01000kg
Quantity 150 x 150mm
 View more versions of this product
Product Code. 15486805
  • 210.18 SEK / Each
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
Monel™ 400 gauze
150 mesh woven from 0.066mm (0.0026 in.) dia. wire
Specifications
Ni:Cu:Fe; 65:33:2 wt%
150 Mesh
150 mesh woven from 0.066mm (0.0026 in.) dia. wire
Monel™ 400 gauze
150 x 150mm