Learn More
Monel|r 400 wire, 0.50mm (0.02in) dia, Thermo Scientific Chemicals
1280.00 SEK - 1630.00 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442127
|
Thermo Scientific Alfa Aesar
045066.TB |
113 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15452127
|
Thermo Scientific Alfa Aesar
045066.TC |
227 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Alloy 400 is widely used in many fields, especially marine and chemical processing. Typical applications are valves and pumps; pump and propeller shafts; marine fixtures and fasteners; electrical and electronic components; springs; chemical processing equipment; gasoline and fresh water tanks; crude petroleum stills, process vessels and piping; boiler feedwater heaters and other heat exchangers; and deaerating heaters.