Läs mer
Monepantel, MedChemExpress
Product Code. 16488757
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
16488757 5 mg -
16482297 50 mg 50mg
16473436 25 mg -
16441439 1 mg -
16406168 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16488757 Supplier Medchem Express Supplier No. HY14774
 Begränsad artikel
Beräknad leverans: 01-09-2026
Add to basket
Add to basket
MedChemExpress Monepantel is organic anthelmintic, and acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits.
Monepantel is organic anthelmintic, and acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits.
Specifications
Kemiskt namn eller material Monepantel
CAS 887148-69-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C20H13F6N3O2S
Kvantitet 5 mg
Synonym AAD1566
Löslighetsinformation DMSO : ≥ 100 mg/mL (211.24 mM)
LEDER O=C(N[C@@](C)(C#N)COC1=CC(C#N)=CC=C1C(F)(F)F)C2=CC=C(SC(F)(F)F)C=C2
Molekylvikt (g/mol) 473.39
Formel vikt 473.39
Procent renhet 99.68%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death